tert-Butyl-3-methylsulphonyloxypiperidine-1-carboxylate 95%
|
|
BD143618-25G
BD143618-5G
BLDPBD143618-25GEA
382
EUR
BLDPBD143618-25G
BLDPBD143618-5G
tert-Butyl-3-methylsulphonyloxypiperidine-1-carboxylate 95%
tert-Butyl-3-methylsulphonyloxypiperidine-1-carboxylate
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₁H₂₁NO₅S Storage Temperature: Refrigerator |
MDL Number:
MFCD07787193 CAS Number: 129888-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...