(Carbobenzoxy)glycylglycine 95%

BD143890-1G BD143890-10G
BLDPBD143890-1GEA 6.3 EUR
BLDPBD143890-1G BLDPBD143890-10G
(Carbobenzoxy)glycylglycine 95%
(Carbobenzoxy)glycylglycine

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₄N₂O₅
MW: 266.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037783
CAS Number: 2566-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR