N-Ethylmaleimide (NEM) 97%
|
|
BD144276-1G
BD144276-100G
BLDPBD144276-1GEA
10.5
EUR
BLDPBD144276-1G
BLDPBD144276-100G
N-Ethylmaleimide (NEM) 97%
N-Ethylmaleimide
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₆H₇NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00005509 CAS Number: 128-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...