2-Iodobiphenyl 98%

BD14445-25G BD14445-10G
BLDPBD14445-25GEA 74.6 EUR
BLDPBD14445-25G BLDPBD14445-10G
2-Iodobiphenyl 98%
2-Iodobiphenyl

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₂H₉I
MW: 280.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039396
CAS Number: 2113-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR