N-tert-Butylmaleimide 97%

BD145751-25G BD145751-100G
BLDPBD145751-25GEA 168 EUR
BLDPBD145751-25G BLDPBD145751-100G
N-tert-Butylmaleimide 97%
N-tert-Butylmaleimide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00191885
CAS Number: 4144-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR