Penciclovir 98%

BD146909-1G BD146909-5G
BLDPBD146909-1GEA 13.7 EUR
BLDPBD146909-1G BLDPBD146909-5G
Penciclovir 98%
Penciclovir

Storage condition: Keep in dark place, Sealed in dry, Store in freezer, under -20°C

Formula: C₁₀H₁₅N₅O₃
Storage Temperature: Freezer
MDL Number: MFCD00866931
CAS Number: 39809-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR