Bis(phenylacetyl)disulphide 97%

BD147624-100G BD147624-10G
BLDPBD147624-100GEA 233 EUR
BLDPBD147624-100G BLDPBD147624-10G
Bis(phenylacetyl)disulphide 97%
Bis(phenylacetyl)disulphide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₆H₁₄O₂S₂
MW: 302.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00513572
CAS Number: 15088-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR