1-Pentyl-4-ethynylbenzene 97%

BD14768-25G BD14768-10G
BLDPBD14768-25GEA 68.3 EUR
BLDPBD14768-25G BLDPBD14768-10G
1-Pentyl-4-ethynylbenzene 97%
1-Pentyl-4-ethynylbenzene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₃H₁₆
Storage Temperature: Ambient
MDL Number: MFCD00173884
CAS Number: 79887-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR