3,4-(Methylenedioxy)phenylboronic acid pinacol ester 95%

BD14832-10G BD14832-25G
BLDPBD14832-10GEA 194 EUR
BLDPBD14832-10G BLDPBD14832-25G
3,4-(Methylenedioxy)phenylboronic acid pinacol ester 95%
3,4-(Methylenedioxy)phenylboronic acid pinacol ester

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₃H₁₇BO₄
MW: 248.0827 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663889
CAS Number: 94838-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR