Poly-(L-lysine) 95%

BD148593-25G BD148593-1G
BLDPBD148593-25GEA 89.3 EUR
BLDPBD148593-25G BLDPBD148593-1G
Poly-(L-lysine) 95%
Poly-(L-lysine)

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: (C₆H₁₄N₂O₂)x
Storage Temperature: Ambient
MDL Number: MFCD01756704
CAS Number: 25104-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR