Potassium methanesulphonate 98%

BD148835-1G BD148835-5G
BLDPBD148835-1GEA 10.5 EUR
BLDPBD148835-1G BLDPBD148835-5G
Potassium methanesulphonate 98%
Potassium methanesulphonate

Storage condition: Inert atmosphere, Room Temperature

Formula: CH₃KO₃S
MW: 134.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070544
CAS Number: 2386-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR