3-((tert-Butoxycarbonyl)amino)-2-hydroxypropanoic acid 97%

BD149191-10G BD149191-1G
BLDPBD149191-10GEA 506 EUR
BLDPBD149191-10G BLDPBD149191-1G
3-((tert-Butoxycarbonyl)amino)-2-hydroxypropanoic acid 97%
3-((tert-Butoxycarbonyl)amino)-2-hydroxypropanoic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₁₅NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD08275906
CAS Number: 218916-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR