Pyrazine-2,5-dicarboxylic acid 97%
|
|
BD149609-500G
BD149609-10G
BLDPBD149609-500GEA
1210
EUR
BLDPBD149609-500G
BLDPBD149609-10G
Pyrazine-2,5-dicarboxylic acid 97%
Pyrazine-2,5-dicarboxylic acid
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₆H₄N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD01630902 CAS Number: 122-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...