Pyridin-4-ylacetonitrile 98%

BD149690-5G BD149690-25G
BLDPBD149690-5GEA 189 EUR
BLDPBD149690-5G BLDPBD149690-25G
Pyridin-4-ylacetonitrile 98%
Pyridin-4-ylacetonitrile

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₆N₂
Storage Temperature: Ambient
MDL Number: MFCD03412601
CAS Number: 13121-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR