5-(Bromomethyl)-2-chloropyridine 98%
|
|
BD149721-1G
BD149721-100MG
BLDPBD149721-1GEA
115
EUR
BLDPBD149721-1G
BLDPBD149721-100MG
5-(Bromomethyl)-2-chloropyridine 98%
5-(Bromomethyl)-2-chloropyridine
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₆H₅BrClN Storage Temperature: Refrigerator |
MDL Number:
MFCD07368893 CAS Number: 182924-36-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...