Pyridinium trifluoromethanesulphonate 98%

BD149780-5G BD149780-100G
BLDPBD149780-5GEA 27.3 EUR
BLDPBD149780-5G BLDPBD149780-100G
Pyridinium trifluoromethanesulphonate 98%
Pyridinium trifluoromethanesulphonate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₆F₃NO₃S
MW: 229.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00274656
CAS Number: 52193-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR