H-Glu(OtBu)-OtBu·HCl 98%

BD14997-100G BD14997-25G BD14997-1G
BLDPBD14997-100GEA 209 EUR
BLDPBD14997-100G BLDPBD14997-25G BLDPBD14997-1G
H-Glu(OtBu)-OtBu·HCl 98%
H-Glu(OtBu)-OtBu·HCl

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₃H₂₆ClNO₄
Storage Temperature: Freezer
MDL Number: MFCD00058003
CAS Number: 32677-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR