Rasagiline mesylate 97%

BD150251-250MG BD150251-25G
BLDPBD150251-250MGEA 15.8 EUR
BLDPBD150251-250MG BLDPBD150251-25G
Rasagiline mesylate 97%
Rasagiline mesylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₃N·CH₄O₃S
MW: 267.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460604
CAS Number: 161735-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR