Roflumilast 99+%

BD150494-1G BD150494-100G
BLDPBD150494-1GEA 36.8 EUR
BLDPBD150494-1G BLDPBD150494-100G
Roflumilast 99+%
Roflumilast

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₇H₁₄Cl₂F₂N₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00938270
CAS Number: 162401-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR