Disodium dihydrogen diphosphate 95%
BD151353-100G
BD151353-1000G
BLDPBD151353-100GEA
9.5
EUR
BLDPBD151353-100G
BLDPBD151353-1000G
Disodium dihydrogen diphosphate 95%
di-Sodium dihydrogen diphosphate
Formula:
H₂Na₂O₇P₂ MW: 221.94 g/mol |
MDL Number:
MFCD00014246 CAS Number: 7758-16-9 Merck Index: 12,08713 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...