4-Ethynyl-1,1'-biphenyl 97%
|
|
BD15144-25G
BD15144-10G
BLDPBD15144-25GEA
265
EUR
BLDPBD15144-25G
BLDPBD15144-10G
4-Ethynyl-1,1'-biphenyl 97%
4-Ethynyl-1,1'-biphenyl
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₁₄H₁₀ Storage Temperature: Ambient |
MDL Number:
MFCD00102191 CAS Number: 29079-00-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...