tert-Butyl 4-bromobenzylcarbamate 98%

BD152922-250MG BD152922-10G
BLDPBD152922-250MGEA 7.4 EUR
BLDPBD152922-250MG BLDPBD152922-10G
tert-Butyl 4-bromobenzylcarbamate 98%
tert-Butyl 4-bromobenzylcarbamate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD08703140
CAS Number: 68819-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR