3-(Benzyloxy)benzaldehyde 98%
|
|
BD15293-500G
BD15293-25G
BLDPBD15293-500GEA
586
EUR
BLDPBD15293-500G
BLDPBD15293-25G
3-(Benzyloxy)benzaldehyde 98%
3-(Benzyloxy)benzaldehyde
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₁₄H₁₂O₂ MW: 212.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003367 CAS Number: 1700-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...