Tetrahydrothiophen-3-one 98%

BD153509-100G BD153509-500G
BLDPBD153509-100GEA 39.9 EUR
BLDPBD153509-100G BLDPBD153509-500G
Tetrahydrothiophen-3-one 98%
Tetrahydrothiophen-3-one

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₄H₆OS
Storage Temperature: Ambient
MDL Number: MFCD00005412
CAS Number: 1003-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR