trans-3-Hexenedioic acid 98%

BD154654-1G BD154654-5G
BLDPBD154654-1GEA 18.9 EUR
BLDPBD154654-1G BLDPBD154654-5G
trans-3-Hexenedioic acid 98%
trans-3-Hexenedioic acid
Formula: C₆H₈O₄
MDL Number: MFCD00002785
CAS Number: 4436-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR