Fmoc-3-(3-pyridyl)-D-alanine 99%

BD1549-1G BD1549-25G
BLDPBD1549-1GEA 60.9 EUR
BLDPBD1549-1G BLDPBD1549-25G
Fmoc-3-(3-pyridyl)-D-alanine 99%
Fmoc-3-(3-pyridyl)-D-alanine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₂₃H₂₀N₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00080267
CAS Number: 142994-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR