1-Thio-β-D-glucose tetraacetate 95%

BD15608-5G BD15608-25G
BLDPBD15608-5GEA 186 EUR
BLDPBD15608-5G BLDPBD15608-25G
1-Thio-β-D-glucose tetraacetate 95%
1-Thio-beta-D-glucose tetraacetate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₄H₂₀O₉S
Storage Temperature: Refrigerator
MDL Number: MFCD00063271
CAS Number: 19879-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR