Voriconazole 99+%

BD156582-25G BD156582-1G
BLDPBD156582-25GEA 1420 EUR
BLDPBD156582-25G BLDPBD156582-1G
Voriconazole 99+%
Voriconazole

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₆H₁₄F₃N₅O
Storage Temperature: Freezer
MDL Number: MFCD00905717
CAS Number: 137234-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR