Cbz-L-tyrosine benzyl ester 97%

BD157566-500G BD157566-25G
BLDPBD157566-500GEA 1210 EUR
BLDPBD157566-500G BLDPBD157566-25G
Cbz-L-tyrosine benzyl ester 97%
Cbz-L-tyrosine benzyl ester

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₄H₂₃NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00136670
CAS Number: 5513-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR