2,3,4-Trifluoro-5-methoxybenzoic acid 98%

BD157665-100MG BD157665-5G
BLDPBD157665-100MGEA 38.9 EUR
BLDPBD157665-100MG BLDPBD157665-5G
2,3,4-Trifluoro-5-methoxybenzoic acid 98%
2,3,4-Trifluoro-5-methoxybenzoic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₅F₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD11973916
CAS Number: 38233-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR