3-Bromocatechol 97%

BD157725-500G BD157725-1G
BLDPBD157725-500GEA 2240 EUR
BLDPBD157725-500G BLDPBD157725-1G
3-Bromocatechol 97%
3-Bromocatechol

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₅BrO₂
Storage Temperature: Ambient
MDL Number: MFCD00869768
CAS Number: 14381-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR