[2-(1,3-Dioxolan-2-yl)ethyl]triphenylphosphonium bromide 98%
|
|
BD1579-100G
BD1579-25G
BLDPBD1579-100GEA
259
EUR
BLDPBD1579-100G
BLDPBD1579-25G
[2-(1,3-Dioxolan-2-yl)ethyl]triphenylphosphonium bromide 98%
[2-(1,3-Dioxolan-2-yl)ethyl]triphenylphosphonium bromide
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₂₃H₂₄BrO₂P Storage Temperature: Refrigerator |
MDL Number:
MFCD00075119 CAS Number: 86608-70-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...