2-Iodobenzo[d]thiazole 95%

BD158121-5G BD158121-250MG BD158121-25G
BLDPBD158121-5GEA 226 EUR
BLDPBD158121-5G BLDPBD158121-250MG BLDPBD158121-25G
2-Iodobenzo[d]thiazole 95%
2-Iodobenzo[d]thiazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₄INS
Storage Temperature: Ambient
MDL Number: MFCD00971971
CAS Number: 1123-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR