2-Bromo-1-chloro-3-fluorobenzene 98%

BD158139-1G BD158139-5G
BLDPBD158139-1GEA 6.3 EUR
BLDPBD158139-1G BLDPBD158139-5G
2-Bromo-1-chloro-3-fluorobenzene 98%
2-Bromo-1-chloro-3-fluorobenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₃BrClF
Storage Temperature: Ambient
MDL Number: MFCD09878161
CAS Number: 309721-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR