3,6-Dihydro-2H-pyran-4-carboxylicacid methyl ester 96%
|
|
BD158441-5G
BD158441-1G
BLDPBD158441-5GEA
740
EUR
BLDPBD158441-5G
BLDPBD158441-1G
3,6-Dihydro-2H-pyran-4-carboxylicacid methyl ester 96%
3,6-Dihydro-2H-pyran-4-carboxylicacid methyl ester
Storage condition: Sealed in dry, 2-8°C
Formula:
C₇H₁₀O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD11706987 CAS Number: 105772-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...