Pyrazolo[1,5-a]pyrimidin-7(4H)-one 98%
|
|
BD158680-100MG
BD158680-25G
BLDPBD158680-100MGEA
32.6
EUR
BLDPBD158680-100MG
BLDPBD158680-25G
Pyrazolo[1,5-a]pyrimidin-7(4H)-one 98%
Pyrazolo[1,5-a]pyrimidin-7(4H)-one
Storage condition: Sealed in dry, Room Temperature
Formula:
C₆H₅N₃O Storage Temperature: Ambient |
MDL Number:
MFCD09746272 CAS Number: 29274-23-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...