7-Bromobenzo[b]thiophene 96%

BD158773-5G BD158773-250MG
BLDPBD158773-5GEA 112 EUR
BLDPBD158773-5G BLDPBD158773-250MG
7-Bromobenzo[b]thiophene 96%
7-Bromobenzo[b]thiophene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₅BrS
Storage Temperature: Ambient
MDL Number: MFCD06657729
CAS Number: 1423-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR