2-Amino-5-bromo-3-pyridinol 98%

BD158879-25G BD158879-250MG
BLDPBD158879-25GEA 81.9 EUR
BLDPBD158879-25G BLDPBD158879-250MG
2-Amino-5-bromo-3-pyridinol 98%
2-Amino-5-bromo-3-pyridinol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₅H₅BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD09744143
CAS Number: 39903-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR