Ethyl-7-bromo-4-hydroxyquinoline-3-carboxylate 95%

BD158996-5G BD158996-1G
BLDPBD158996-5GEA 112 EUR
BLDPBD158996-5G BLDPBD158996-1G
Ethyl-7-bromo-4-hydroxyquinoline-3-carboxylate 95%
Ethyl 7-bromo-4-hydroxyquinoline-3-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₂H₁₀BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD00487280
CAS Number: 179943-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR