5-Bromopyrimidin-4(3H)-one sulphate(2:1) 95%

BD159750-25G BD159750-1G
BLDPBD159750-25GEA 343 EUR
BLDPBD159750-25G BLDPBD159750-1G
5-Bromopyrimidin-4(3H)-one sulphate(2:1) 95%
5-Bromopyrimidin-4(3H)-one sulphate(2:1)

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₈Br₂N₄O₆S
Storage Temperature: Ambient
MDL Number: MFCD13195360
CAS Number: 97234-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR