2-Chloro-(5-methoxycarbonyl)phenylboronic acid 96%

BD159817-1G BD159817-5G
BLDPBD159817-1GEA 110 EUR
BLDPBD159817-1G BLDPBD159817-5G
2-Chloro-(5-methoxycarbonyl)phenylboronic acid 96%
2-Chloro-(5-methoxycarbonyl)phenylboronic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₈BClO₄
Storage Temperature: Refrigerator
MDL Number: MFCD08689528
CAS Number: 913835-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR