Phenethyl-p-toluenesulphonate 98%

BD15987-5G BD15987-100G
BLDPBD15987-5GEA 23.1 EUR
BLDPBD15987-5G BLDPBD15987-100G
Phenethyl-p-toluenesulphonate 98%
Phenethyl-p-toluenesulphonate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₅H₁₆O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00059322
CAS Number: 4455-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR