tert-Butyl-3-phenylpiperazine-1-carboxylate 98%
|
|
BD16134-25G
BD16134-100MG
BD16134-250MG
BLDPBD16134-25GEA
1530
EUR
BLDPBD16134-25G
BLDPBD16134-100MG
BLDPBD16134-250MG
tert-Butyl-3-phenylpiperazine-1-carboxylate 98%
tert-Butyl-3-phenylpiperazine-1-carboxylate
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formula:
C₁₅H₂₂N₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD03840082 CAS Number: 502649-25-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...