6-Bromophthalazin-1(4H)-one 98%

BD161559-1G BD161559-100G BD161559-5G
BLDPBD161559-1GEA 33.6 EUR
BLDPBD161559-1G BLDPBD161559-100G BLDPBD161559-5G
6-Bromophthalazin-1(4H)-one 98%
6-Bromophthalazin-1(4H)-one

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₅BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD09264003
CAS Number: 75884-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR