Methyl-5-methylthiophene-3-carboxylate 97%

BD161955-1G BD161955-5G
BLDPBD161955-1GEA 158 EUR
BLDPBD161955-1G BLDPBD161955-5G
Methyl-5-methylthiophene-3-carboxylate 97%
Methyl-5-methylthiophene-3-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₇H₈O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD02245126
CAS Number: 88770-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR