2-Amino-5-bromo-4-fluorobenzoic acid 97%

BD162116-100G BD162116-10G
BLDPBD162116-100GEA 461 EUR
BLDPBD162116-100G BLDPBD162116-10G
2-Amino-5-bromo-4-fluorobenzoic acid 97%
2-Amino-5-bromo-4-fluorobenzoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₅BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD06409112
CAS Number: 143945-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR