Fmoc-3-bromo-D-phenylalanine 98%

BD1626-5G BD1626-25G
BLDPBD1626-5GEA 284 EUR
BLDPBD1626-5G BLDPBD1626-25G
Fmoc-3-bromo-D-phenylalanine 98%
Fmoc-3-bromo-D-phenylalanine

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₄H₂₀BrNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD01311771
CAS Number: 220497-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR