Ethyl 6-amino-1H-indole-7-carboxylate 95%
|
|
BD163652-250MG
BD163652-1G
BLDPBD163652-250MGEA
219
EUR
BLDPBD163652-250MG
BLDPBD163652-1G
Ethyl 6-amino-1H-indole-7-carboxylate 95%
Ethyl 6-amino-1H-indole-7-carboxylate
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₁₁H₁₂N₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD13195348 CAS Number: 174311-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...