7,8-Dimethoxy-3-(3-iodopropyl)-1,3-dihydro-2H-3-benzazepin-2-one 95%

BD163955-1G BD163955-5G
BLDPBD163955-1GEA 9.5 EUR
BLDPBD163955-1G BLDPBD163955-5G
7,8-Dimethoxy-3-(3-iodopropyl)-1,3-dihydro-2H-3-benzazepin-2-one 95%
7,8-Dimethoxy-3-(3-iodopropyl)-1,3-dihydro-2H-3-benzazepin-2-one

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₅H₁₈INO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09955403
CAS Number: 148870-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR