Methyl 5-hydroxy-2-methylbenzoate 98%

BD164098-100MG BD164098-25G
BLDPBD164098-100MGEA 15.8 EUR
BLDPBD164098-100MG BLDPBD164098-25G
Methyl 5-hydroxy-2-methylbenzoate 98%
Methyl-5-hydroxy-2-methylbenzoate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226199
CAS Number: 73505-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR